siRNA Details
| Gene Symbol: Ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013863.2 | NP_001013885.2 | 5 | 1010 | ||
| XM_008768859.1 | XP_008767081.1 | 6 | 1043 | |||
| XM_008768860.1 | XP_008767082.1 | 7 | 1459 | |||
| XM_008768861.1 | XP_008767083.1 | 7 | 1208 | |||
| GenBank mRNA | BC086565.1 | - | 5 | 1160 | 7113479 | |

Ydjc
Genomic Map