siRNA Details
Gene Symbol: Crkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008284.1 | NP_001008285.1 | 1192 | |||
GenBank mRNA | BC085865.1 | - | 1192 | 7190712 |
Gene Symbol: LOC100911248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751095.3 | XP_003751143.1 | 2 | 1224 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
