siRNA Details
| Gene Symbol: Scarf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105864.1 | NP_001099334.1 | 3 | 328 | ||
| XM_006248688.2 | XP_006248750.1 | 3 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scarf2| Gene Symbol: Scarf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105864.1 | NP_001099334.1 | 3 | 328 | ||
| XM_006248688.2 | XP_006248750.1 | 3 | 316 | |||