siRNA Details
| Gene Symbol: Dgcr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105865.1 | NP_001099335.1 | 3 | 1293 | ||
| XM_006248606.2 | XP_006248668.1 | 2 | 1120 | |||
| XM_006248607.2 | XP_006248669.1 | 2 | 4819 | |||
| XM_008768865.1 | XP_008767087.1 | 1 | 5222 | |||

Dgcr8
Genomic Map