siRNA Details
| Gene Symbol: Dgcr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105865.1 | NP_001099335.1 | 11 | 2431 | ||
| XM_006248606.2 | XP_006248668.1 | 10 | 2258 | |||
| XM_006248607.2 | XP_006248669.1 | 10 | 5957 | |||
| XM_008768865.1 | XP_008767087.1 | 9 | 6360 | |||

Dgcr8
Genomic Map