siRNA Details
| Gene Symbol: Mrpl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024865.1 | NP_001020036.1 | 3 | 285 | ||
| GenBank mRNA | BC097472.1 | - | 3 | 285 | 7457772 | |
| Gene Symbol: LOC684557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252309.2 | XP_006252371.1 | 1 | 69 | ||

Mrpl40
Genomic Map