siRNA Details
| Gene Symbol: Olr1567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000043.1 | NP_001000043.1 | 497 | |||
| Gene Symbol: LOC100910999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751087.2 | XP_003751135.1 | 1 | 497 | ||

Olr1567
Genomic Map