siRNA Details
| Gene Symbol: Olr1566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000044.1 | NP_001000044.1 | 1 | 306 | ||
| Gene Symbol: Olr1565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000908.1 | NP_001000908.1 | 1 | 309 | ||

Olr1566
Genomic Map