siRNA Details
| Gene Symbol: Klhl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105867.1 | NP_001099337.1 | 4 | 1012 | ||
| XM_006248594.1 | XP_006248656.1 | 4 | 783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl6| Gene Symbol: Klhl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105867.1 | NP_001099337.1 | 4 | 1012 | ||
| XM_006248594.1 | XP_006248656.1 | 4 | 783 | |||