siRNA Details
| Gene Symbol: Parl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035249.1 | NP_001030326.1 | 10 | 1135 | ||
| GenBank mRNA | BC088226.1 | - | 10 | 1053 | ||
| BC105907.1 | - | 10 | 1135 | |||
| BC126098.1 | - | 10 | 1167 | |||
| BC158564.1 | - | 10 | 1119 | 8374563 | ||
| FQ214115.1 | - | 10 | 1130 | |||

Parl
Genomic Map