siRNA Details
| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011896.1 | NP_001011896.1 | 4, 5 | 439 | ||
| GenBank mRNA | BC082042.1 | - | 4, 5 | 439 | 7131554 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcf3| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011896.1 | NP_001011896.1 | 4, 5 | 439 | ||
| GenBank mRNA | BC082042.1 | - | 4, 5 | 439 | 7131554 | |