siRNA Details
| Gene Symbol: Vps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248575.2 | XP_006248637.1 | 44 | 4028 | ||
| XM_006248577.2 | XP_006248639.1 | 44 | 4020 | |||
| XM_006248578.2 | XP_006248640.1 | 44 | 4052 | |||
| XM_006248579.2 | XP_006248641.1 | 43 | 4022 | |||
| XM_008768830.1 | XP_008767052.1 | 44 | 4227 | |||
| XM_008768831.1 | XP_008767053.1 | 44 | 4028 | |||

Vps8
Genomic Map