siRNA Details
Gene Symbol: Kng1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009628.1 | NP_001009628.1 | 1 | 228 | ||
GenBank mRNA | BC088161.1 | - | 1 | 228 | 7369906 | |
M11661.1 | - | 1 | 126 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kng1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009628.1 | NP_001009628.1 | 1 | 228 | ||
GenBank mRNA | BC088161.1 | - | 1 | 228 | 7369906 | |
M11661.1 | - | 1 | 126 |