siRNA Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182736.1 | NP_877400.1 | 10 | 857 | ||
| XM_008768795.1 | XP_008767017.1 | 11 | 1622 | |||
| GenBank mRNA | AJ534984.1 | - | 10 | 857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc50| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182736.1 | NP_877400.1 | 10 | 857 | ||
| XM_008768795.1 | XP_008767017.1 | 11 | 1622 | |||
| GenBank mRNA | AJ534984.1 | - | 10 | 857 | ||