siRNA Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013421.1 | NP_001013439.1 | 12, 13 | 1625 | ||
| GenBank mRNA | BC079013.1 | - | 11, 12 | 1471 | ||
| BC091338.1 | - | 12, 13 | 1625 | 7315079 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsg1| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013421.1 | NP_001013439.1 | 12, 13 | 1625 | ||
| GenBank mRNA | BC079013.1 | - | 11, 12 | 1471 | ||
| BC091338.1 | - | 12, 13 | 1625 | 7315079 | ||