siRNA Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013421.1 | NP_001013439.1 | 1, 2 | 125 | ||
| GenBank mRNA | AF452725.1 | - | 74 | |||
| AF452729.1 | - | 74 | ||||
| BC079013.1 | - | 1, 2 | 124 | |||
| BC091338.1 | - | 1, 2 | 125 | 7315079 | ||

Lsg1
Genomic Map