siRNA Details
| Gene Symbol: Ropn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025628.1 | NP_001020799.1 | 5 | 716 | ||
| XM_006248431.2 | XP_006248493.1 | 5 | 659 | |||
| GenBank mRNA | BC099132.1 | - | 5 | 716 | 7453834 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ropn1| Gene Symbol: Ropn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025628.1 | NP_001020799.1 | 5 | 716 | ||
| XM_006248431.2 | XP_006248493.1 | 5 | 659 | |||
| GenBank mRNA | BC099132.1 | - | 5 | 716 | 7453834 | |