siRNA Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198726.2 | NP_942021.2 | 1 | 100 | ||
| XM_008768713.1 | XP_008766935.1 | 2 | 300 | |||
| GenBank mRNA | AY351984.1 | - | 1 | 169 | ||
| AY779025.1 | - | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kpna1| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198726.2 | NP_942021.2 | 1 | 100 | ||
| XM_008768713.1 | XP_008766935.1 | 2 | 300 | |||
| GenBank mRNA | AY351984.1 | - | 1 | 169 | ||
| AY779025.1 | - | 1 | 100 | |||