siRNA Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105875.1 | NP_001099345.1 | 3, 4 | 865 | ||
| GenBank mRNA | FQ221021.1 | - | 3, 4 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc58| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105875.1 | NP_001099345.1 | 3, 4 | 865 | ||
| GenBank mRNA | FQ221021.1 | - | 3, 4 | 328 | ||