siRNA Details
| Gene Symbol: Ndufb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037338.1 | NP_001032415.1 | 3 | 354 | ||
| GenBank mRNA | CB732095.1 | - | 3 | 354 | ||
| DQ255901.1 | - | 3 | 207 | |||
| FQ217067.1 | - | 3 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb4| Gene Symbol: Ndufb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037338.1 | NP_001032415.1 | 3 | 354 | ||
| GenBank mRNA | CB732095.1 | - | 3 | 354 | ||
| DQ255901.1 | - | 3 | 207 | |||
| FQ217067.1 | - | 3 | 361 | |||