siRNA Details
| Gene Symbol: RGD1566314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248319.2 | XP_006248381.1 | 9 | 1424 | ||
| XM_008768763.1 | XP_008766985.1 | 9 | 1424 | |||
| XM_008768764.1 | XP_008766986.1 | 9 | 1424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1566314| Gene Symbol: RGD1566314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248319.2 | XP_006248381.1 | 9 | 1424 | ||
| XM_008768763.1 | XP_008766985.1 | 9 | 1424 | |||
| XM_008768764.1 | XP_008766986.1 | 9 | 1424 | |||