siRNA Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100653.1 | NP_001094123.1 | 19 | 1962 | ||
| XM_006248328.2 | XP_006248390.1 | 19 | 2844 | |||
| XM_008768718.1 | XP_008766940.1 | 19 | 2843 | |||
| GenBank mRNA | AY562215.1 | - | 19 | 1962 | ||

Sidt1
Genomic Map