siRNA Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100653.1 | NP_001094123.1 | 8 | 877 | ||
| XM_006248328.2 | XP_006248390.1 | 8 | 1759 | |||
| XM_008768718.1 | XP_008766940.1 | 8 | 1758 | |||
| GenBank mRNA | AY562215.1 | - | 8 | 877 | ||

Sidt1
Genomic Map