siRNA Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008285.1 | NP_001008286.1 | 11 | 1237 | ||
| XM_006248331.2 | XP_006248393.1 | 12 | 1451 | |||
| XM_006248332.2 | XP_006248394.1 | 11 | 1405 | |||
| XM_006248334.1 | XP_006248396.1 | 9 | 1134 | |||
| XM_008768719.1 | XP_008766941.1 | 11 | 1335 | |||
| XM_008768721.1 | XP_008766943.1 | 9 | 1666 | |||
| GenBank mRNA | BC085954.1 | - | 11 | 1237 | 7134530 | |
| FQ210155.1 | - | 11 | 1303 | |||

Spice1
Genomic Map