siRNA Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008285.1 | NP_001008286.1 | 6 | 494 | ||
| XM_006248331.2 | XP_006248393.1 | 7 | 708 | |||
| XM_006248332.2 | XP_006248394.1 | 6 | 662 | |||
| XM_006248334.1 | XP_006248396.1 | 4 | 391 | |||
| XM_008768719.1 | XP_008766941.1 | 6 | 592 | |||
| XM_008768721.1 | XP_008766943.1 | 4 | 923 | |||
| GenBank mRNA | BC085954.1 | - | 6 | 494 | 7134530 | |
| FQ210155.1 | - | 6 | 560 | |||

Spice1
Genomic Map