siRNA Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105883.1 | NP_001099353.1 | 2 | 414 | ||
| XM_006248335.2 | XP_006248397.1 | 2 | 452 | |||
| XM_006248337.1 | XP_006248399.1 | 2 | 452 | |||
| XM_006248338.2 | XP_006248400.1 | 2 | 452 | |||
| XM_008768723.1 | XP_008766945.1 | 2 | 395 | |||
| XM_008768724.1 | XP_008766946.1 | 2 | 1109 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021497, NM_021496, NM_021495, AF195833, AK011949, and others.

Pvrl3
Genomic Map