siRNA Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248335.2 | XP_006248397.1 | 6 | 1489 | ||
| XM_006248337.1 | XP_006248399.1 | 6 | 1489 | |||
| XM_008768723.1 | XP_008766945.1 | 6 | 1432 | |||
| XM_008768724.1 | XP_008766946.1 | 6 | 2146 | |||
| GenBank mRNA | FQ221474.1 | - | 1 | 93 | ||

Pvrl3
Genomic Map