siRNA Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105883.1 | NP_001099353.1 | 3 | 658 | ||
| XM_006248335.2 | XP_006248397.1 | 3 | 696 | |||
| XM_006248337.1 | XP_006248399.1 | 3 | 696 | |||
| XM_006248338.2 | XP_006248400.1 | 3 | 696 | |||
| XM_008768723.1 | XP_008766945.1 | 3 | 639 | |||
| XM_008768724.1 | XP_008766946.1 | 3 | 1353 | |||

Pvrl3
Genomic Map