siRNA Details
| Gene Symbol: Retnlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181625.1 | NP_853656.1 | 2 | 142 | ||
| XM_006248282.2 | XP_006248344.1 | 3 | 235 | |||
| GenBank mRNA | AJ514932.1 | - | 2 | 142 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Retnlg| Gene Symbol: Retnlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181625.1 | NP_853656.1 | 2 | 142 | ||
| XM_006248282.2 | XP_006248344.1 | 3 | 235 | |||
| GenBank mRNA | AJ514932.1 | - | 2 | 142 | ||