siRNA Details
| Gene Symbol: RGD1565996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768672.1 | XP_008766894.1 | 1 | 157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565996| Gene Symbol: RGD1565996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768672.1 | XP_008766894.1 | 1 | 157 | ||