siRNA Details
| Gene Symbol: Hoxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105885.1 | NP_001099355.1 | 6 | 2361 | ||
| XM_008761882.1 | XP_008760104.1 | 3 | 7069 | |||
| XM_008761883.1 | XP_008760105.1 | 3 | 5796 | |||
| XM_008761884.1 | XP_008760106.1 | 4 | 6548 | |||
| XM_008761885.1 | XP_008760107.1 | 4 | 6970 | |||
| XM_008761886.1 | XP_008760108.1 | 4 | 4983 | |||
| XM_008761887.1 | XP_008760109.1 | 5 | 4884 | |||

Hoxd4
Genomic Map