siRNA Details
| Gene Symbol: Pcnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047846.1 | NP_001041311.1 | 6 | 542 | ||
| XM_006248241.2 | XP_006248303.1 | 5 | 515 | |||
| XM_006248242.1 | XP_006248304.1 | 5 | 513 | |||
| XM_008768630.1 | XP_008766852.1 | 5 | 574 | |||
| XM_008768631.1 | XP_008766853.1 | 5 | 571 | |||
| GenBank mRNA | AY325209.1 | - | 6 | 542 | ||

Pcnp
Genomic Map