siRNA Details
| Gene Symbol: Senp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105888.1 | NP_001099358.1 | 6 | 613 | ||
| XM_006248244.2 | XP_006248306.1 | 6 | 1568 | |||
| XM_006248247.2 | XP_006248309.1 | 5 | 1371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Senp7| Gene Symbol: Senp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105888.1 | NP_001099358.1 | 6 | 613 | ||
| XM_006248244.2 | XP_006248306.1 | 6 | 1568 | |||
| XM_006248247.2 | XP_006248309.1 | 5 | 1371 | |||