siRNA Details
| Gene Symbol: Olr1549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000052.1 | NP_001000052.1 | 1 | 237 | ||
| Gene Symbol: LOC103693566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768681.1 | XP_008766903.1 | 1 | 237 | ||
| Gene Symbol: Olr1548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001108.1 | NP_001001108.1 | 1 | 237 | ||

Olr1549
Genomic Map