siRNA Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002802.1 | NP_001002802.1 | 5 | 806 | ||
| XM_006248153.2 | XP_006248215.1 | 5 | 860 | |||
| GenBank mRNA | BC166597.1 | - | 5 | 820 | 7936295 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bace2| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002802.1 | NP_001002802.1 | 5 | 806 | ||
| XM_006248153.2 | XP_006248215.1 | 5 | 860 | |||
| GenBank mRNA | BC166597.1 | - | 5 | 820 | 7936295 | |