siRNA Details
Gene Symbol: Wrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199373.1 | NP_955405.1 | 3 | 389 | ||
GenBank mRNA | BC062049.1 | - | 3 | 389 | 5598163 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199373.1 | NP_955405.1 | 3 | 389 | ||
GenBank mRNA | BC062049.1 | - | 3 | 389 | 5598163 |