siRNA Details
| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105891.1 | NP_001099361.1 | 4 | 711 | ||
| XM_006248154.2 | XP_006248216.1 | 5 | 596 | |||
| XM_008768571.1 | XP_008766793.1 | 5 | 868 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmg1| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105891.1 | NP_001099361.1 | 4 | 711 | ||
| XM_006248154.2 | XP_006248216.1 | 5 | 596 | |||
| XM_008768571.1 | XP_008766793.1 | 5 | 868 | |||