siRNA Details
| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024741.1 | NP_001019912.1 | 6 | 659 | ||
| XM_006248044.2 | XP_006248106.1 | 6 | 695 | |||
| GenBank mRNA | BC097460.1 | - | 6 | 659 | 7457454 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chaf1b| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024741.1 | NP_001019912.1 | 6 | 659 | ||
| XM_006248044.2 | XP_006248106.1 | 6 | 695 | |||
| GenBank mRNA | BC097460.1 | - | 6 | 659 | 7457454 | |