siRNA Details
| Gene Symbol: Ifitm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_221637.1 | XP_221637.1 | 1 | 355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm7| Gene Symbol: Ifitm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_221637.1 | XP_221637.1 | 1 | 355 | ||