siRNA Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013044.1 | NP_001013062.1 | 9, 10 | 718 | ||
| XM_006248045.2 | XP_006248107.1 | 7, 8 | 1220 | |||
| XM_006248046.2 | XP_006248108.1 | 8, 9 | 651 | |||
| GenBank mRNA | BC083853.1 | - | 9, 10 | 718 | 7124012 | |

Cryzl1
Genomic Map