siRNA Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013044.1 | NP_001013062.1 | 6, 7 | 364 | ||
| XM_006248046.2 | XP_006248108.1 | 5, 6 | 297 | |||
| GenBank mRNA | BC083853.1 | - | 6, 7 | 364 | 7124012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryzl1| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013044.1 | NP_001013062.1 | 6, 7 | 364 | ||
| XM_006248046.2 | XP_006248108.1 | 5, 6 | 297 | |||
| GenBank mRNA | BC083853.1 | - | 6, 7 | 364 | 7124012 | |