siRNA Details
| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011899.2 | NP_001011899.2 | 7 | 760 | ||
| GenBank mRNA | BC087644.1 | - | 7 | 722 | ||
| BC168661.1 | - | 7 | 749 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gart| Gene Symbol: Gart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011899.2 | NP_001011899.2 | 7 | 760 | ||
| GenBank mRNA | BC087644.1 | - | 7 | 722 | ||
| BC168661.1 | - | 7 | 749 | |||