siRNA Details
| Gene Symbol: Hunk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191662.1 | NP_001178591.1 | 10 | 1334 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hunk| Gene Symbol: Hunk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191662.1 | NP_001178591.1 | 10 | 1334 | ||