siRNA Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100502.1 | NP_001093972.1 | 12 | 1830 | ||
| GenBank mRNA | BC090037.1 | - | 9 | 1205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbe1| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100502.1 | NP_001093972.1 | 12 | 1830 | ||
| GenBank mRNA | BC090037.1 | - | 9 | 1205 | ||