siRNA Details
| Gene Symbol: Cd209b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170397.1 | NP_001163868.1 | 10 | 864 | ||
| XM_008768946.1 | XP_008767168.1 | 10 | 945 | |||
| GenBank mRNA | BC091252.1 | - | 10 | 864 | ||
| FQ227772.1 | - | 10 | 870 | |||

Cd209b
Genomic Map