siRNA Details
Gene Symbol: LOC100363914 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769020.1 | XP_008767242.1 | 2 | 74 |
Gene Symbol: LOC100911529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769019.1 | XP_008767241.1 | 1 | 74 |
