siRNA Details
| Gene Symbol: Rfc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009629.1 | NP_001009629.1 | 6, 7 | 779 | ||
| XM_006248734.2 | XP_006248796.1 | 5, 6 | 670 | |||
| GenBank mRNA | BC088281.1 | - | 6, 7 | 779 | 7383278 | |
| FQ216316.1 | - | 6, 7 | 768 | |||
| FQ232102.1 | - | 6, 7 | 751 | |||

Rfc3
Genomic Map