siRNA Details
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105906.2 | NP_001099376.1 | 7 | 1330 | ||
NM_001198555.1 | NP_001185484.1 | 7 | 1330 | |||
XM_006248810.2 | XP_006248872.1 | 6 | 1273 | |||
GenBank mRNA | CO396482.1 | - | 3 | 243 | ||
EV774129.1 | - | 2 | 138 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
