siRNA Details
| Gene Symbol: Uspl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105906.2 | NP_001099376.1 | 7 | 1361 | ||
| NM_001198555.1 | NP_001185484.1 | 7 | 1361 | |||
| XM_006248810.2 | XP_006248872.1 | 6 | 1304 | |||
| GenBank mRNA | CO396482.1 | - | 3 | 274 | ||
| EV774129.1 | - | 2 | 169 | |||

Uspl1
Genomic Map