siRNA Details
| Gene Symbol: Pomp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100942.1 | NP_001094412.1 | 5, 6 | 371 | ||
| GenBank mRNA | BC161800.1 | - | 5, 6 | 414 | 9026464 | |
| EF535528.1 | - | 5, 6 | 371 | |||
| FQ217734.1 | - | 4, 5 | 362 | |||
| FQ221470.1 | - | 5, 6 | 388 | |||
| FQ224264.1 | - | 5, 6 | 408 | |||
| Gene Symbol: LOC100911238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248840.1 | XP_006248902.1 | 5, 6 | 458 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025624, AK003439, AK011115, AK013934, AK132041, and others.

Pomp
Genomic Map